Merck
Synthesis of taxoids with improved cytotoxicity and solubility for use in tumor-specific delivery.
Journal of medicinal chemistry (2004-09-17)
Michael L Miller, Elizabeth E Roller, Robert Y Zhao, Barbara A Leece, Olga Ab, Erkan Baloglu, Victor S Goldmacher, Ravi V J Chari
ABSTRACT
To develop effective taxane-antibody immunoconjugates, we have prepared a series of modified taxanes that have both improved toxicity and solubility in aqueous systems as compared to paclitaxel (1a). These taxanes have been modified at either the C-10 or C-7 position and were found to be very cytotoxic against both normal and multi-drug-resistant (MDR) cells, as well as up to 30 times more soluble than paclitaxel in various buffer systems.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Hydroxy-PEG4-t-butyl ester