Merck
  • KIF4A facilitates cell proliferation via induction of p21-mediated cell cycle progression and promotes metastasis in colorectal cancer.
KIF4A facilitates cell proliferation via induction of p21-mediated cell cycle progression and promotes metastasis in colorectal cancer.
Cell death & disease (2018-05-01)
Ping-Fu Hou, Tao Jiang, Fang Chen, Pei-Cong Shi, Hai-Qing Li, Jin Bai, Jun Song
ABSTRACT
Kinesin family member 4A (KIF4A) was found to be implicated in the regulation of chromosome condensation and segregation during mitotic cell division, which is essential for eukaryotic cell proliferation. However, little is known about the role of KIF4A in colorectal carcinoma (CRC). This study explored the biological function of KIF4A in CRC progression and investigated the potential molecular mechanisms involved. Here, we found that KIF4A was remarkably upregulated in primary CRC tissues and cell lines compared with paired non-cancerous tissues and normal colorectal epithelium. Elevated expression of KIF4A in CRC tissues was significantly correlated with clinicopathological characteristics in patients as well as with shorter overall and disease-free cumulative survival. Multivariate Cox regression analysis revealed that KIF4A was an independent prognostic factor for poor survival in human CRC patients. Functional assays, including a CCK-8 cell proliferation assay, colony formation analysis, cancer xenografts in nude mice, cell cycle and apoptosis analysis, indicated that KIF4A obviously enhanced cell proliferation by promoting cell cycle progression in vitro and in vivo. Furthermore, gene set enrichment analysis, Luciferase reporter assays, and ChIP assays revealed that KIF4A facilitates cell proliferation via regulating the p21 promoter, whereas KIF4A had no effect on cell apoptosis. In addition, Transwell analysis indicated that KIF4A promotes migration and invasion in CRC. Taken together, these findings not only demonstrate that KIF4A contributes to CRC proliferation via modulation of p21-mediated cell cycle progression but also suggest the potential value of KIF4A as a clinical prognostic marker and target for molecular treatments.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
MISSION® esiRNA, targeting human KIF4A