Merck
  • Mesenchymal stromal cells from human umbilical cord prevent the development of lung fibrosis in immunocompetent mice.
Mesenchymal stromal cells from human umbilical cord prevent the development of lung fibrosis in immunocompetent mice.
PloS one (2018-06-02)
Gianluca Moroncini, Chiara Paolini, Fiorenza Orlando, Chiara Capelli, Antonella Grieco, Cecilia Tonnini, Silvia Agarbati, Eleonora Mondini, Stefania Saccomanno, Gaia Goteri, Silvia Svegliati Baroni, Mauro Provinciali, Martino Introna, Nicoletta Del Papa, Armando Gabrielli
ABSTRACT
Lung fibrosis is a severe condition resulting from several interstial lung diseases (ILD) with different etiologies. Current therapy of ILD, especially those associated with connective tissue diseases, is rather limited and new anti-fibrotic strategies are needed. In this study, we investigated the anti-fibrotic activity in vivo of human mesenchymal stromal cells obtained from whole umbilical cord (hUC-MSC). Adult immunocompetent C57BL/6 mice (n. = 8 for each experimental condition) were injected intravenously with hUC-MSC (n. = 2.5 × 105) twice, 24 hours and 7 days after endotracheal injection of bleomycin. Upon sacrifice at days 8, 14, 21, collagen content, inflammatory cytokine profile, and hUC-MSC presence in explanted lung tissue were analyzed. Systemic administration of a double dose of hUC-MSC significantly reduced bleomycin-induced lung injury (inflammation and fibrosis) in mice through a selective inhibition of the IL6-IL10-TGFβ axis involving lung M2 macrophages. Only few hUC-MSC were detected from explanted lungs, suggesting a "hit and run" mechanism of action of this cellular therapy. Our data indicate that hUC-MSC possess strong in vivo anti-fibrotic activity in a mouse model resembling an immunocompetent human subject affected by inflammatory ILD, providing proof of concept for ad-hoc clinical trials.