Merck
PSMD5 Inactivation Promotes 26S Proteasome Assembly during Colorectal Tumor Progression.
Cancer research (2018-05-03)
Avi Levin, Adi Minis, Gadi Lalazar, Jose Rodriguez, Hermann Steller
ABSTRACT
Protein degradation by the ubiquitin-proteasome system (UPS) is central to protein homeostasis and cell survival. The active 26S proteasome is a large protease complex consisting of a catalytic 20S subunit and 19S regulatory particles. Cancer cells are exposed to considerable protein overload due to high metabolic rates, reprogrammed energy metabolism, and aneuploidy. Here we report a mechanism that facilitates the assembly of active 26S proteasomes in malignant cells. Upon tumorigenic transformation of the gut epithelium, 26S proteasome assembly was significantly enhanced, but levels of individual subunits were not changed. This enhanced assembly of 26S proteasomes increased further with tumor progression and was observed specifically in transformed cells, but not in other rapidly dividing cells. Moreover, expression of PSMD5, an inhibitor of proteasome assembly, was reduced in intestinal tumors and silenced with tumor progression. Reexpression of PSMD5 in tumor cells caused decreased 26S assembly and accumulation of polyubiquitinated proteins. These results suggest that inhibition of cancer-associated proteasome assembly may provide a novel therapeutic strategy to selectively kill cancer cells.Significance: Enhanced cancer-associated proteasome assembly is a major stress response that allows tumors to adapt to and to withstand protein overload.Graphical Abstract: http://cancerres.aacrjournals.org/content/canres/78/13/3458/F1.large.jpg Cancer Res; 78(13); 3458-68. ©2018 AACR.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)
Sigma-Aldrich
MISSION® esiRNA, targeting human PSMD5