Merck
HOXA13 directly regulates EphA6 and EphA7 expression in the genital tubercle vascular endothelia.
Developmental dynamics : an official publication of the American Association of Anatomists (2007-02-17)
Carley A Shaut, Chie Saneyoshi, Emily A Morgan, Wendy M Knosp, Diane R Sexton, H Scott Stadler
ABSTRACT
Hypospadias, a common defect affecting the growth and closure of the external genitalia, is often accompanied by gross enlargements of the genital tubercle (GT) vasculature. Because Hoxa13 homozygous mutant mice also exhibit hypospadias and GT vessel expansion, we examined whether genes playing a role in angiogenesis exhibit reduced expression in the GT. From this analysis, reductions in EphA6 and EphA7 were detected. Characterization of EphA6 and EphA7 expression in the GT confirmed colocalization with HOXA13 in the GT vascular endothelia. Analysis of the EphA6 and EphA7 promoter regions revealed a series of highly conserved cis-regulatory elements bound by HOXA13 with high affinity. GT chromatin immunoprecipitation confirmed that HOXA13 binds these gene-regulatory elements in vivo. In vitro, HOXA13 activates gene expression through the EphA6 and EphA7 gene-regulatory elements. Together these findings indicate that HOXA13 directly regulates EphA6 and EphA7 in the developing GT and identifies the GT vascular endothelia as a novel site for HOXA13-dependent expression of EphA6 and EphA7.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
EPHA6 (561-end), active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution