Merck
  • Enzyme biosensor for androsterone based on 3alpha-hydroxysteroid dehydrogenase immobilized onto a carbon nanotubes/ionic liquid/NAD+ composite electrode
Enzyme biosensor for androsterone based on 3alpha-hydroxysteroid dehydrogenase immobilized onto a carbon nanotubes/ionic liquid/NAD+ composite electrode
Talanta (2012)
Mundaca RA, et al.