Merck
  • Phosphazene Superbase-Mediated Regio- and Stereoselective Iodoaminocyclization of 2-(1-Alkynyl)benzamides for the Synthesis of Isoindolin-1-ones.
Phosphazene Superbase-Mediated Regio- and Stereoselective Iodoaminocyclization of 2-(1-Alkynyl)benzamides for the Synthesis of Isoindolin-1-ones.
The Journal of organic chemistry (2019-04-16)
Saurabh Mehta, Dhirendra Brahmchari
ABSTRACT
Phosphazene superbase P4- t-Bu mediated iodoaminocyclization of 2-(1-alkynyl)benzamides is reported. The reaction works under ambient conditions and instantaneously results in the synthesis of isoindolin-1-ones in 65-97% yields, in a regio- and stereoselective manner. The exclusive formation of products with Z-geometry (across the exo C═C bond) has been confirmed through X-ray crystallography. The methodology also provides an easy access to aristolactams, an important class of natural products. This has been successfully demonstrated by synthesizing two aristolactam derivatives (including Cepharanone B).
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Phosphazene base P4-t-Bu solution, ~0.8 M in hexane