Merck
  • Purification and characterization of human kallikrein 11, a candidate prostate and ovarian cancer biomarker, from seminal plasma.
Purification and characterization of human kallikrein 11, a candidate prostate and ovarian cancer biomarker, from seminal plasma.
Clinical cancer research : an official journal of the American Association for Cancer Research (2006-02-10)
Liu-Ying Luo, Shannon J C Shan, Marc B Elliott, Antoninus Soosaipillai, Eleftherios P Diamandis
ABSTRACT
Preliminary data suggest that hK11 is a novel serum biomarker for prostate and ovarian cancer. To examine the enzymatic characteristics of hK11, we purified and functionally characterized native hK11 from seminal plasma. hK11 was purified from seminal plasma by immunoaffinity chromatography and characterized by kinetic analysis, electrophoresis, Western blots, and mass spectrometry. hK11 is present in seminal plasma at concentrations ranging from 2 to 37 microg/mL. Using immunoaffinity chromatography and reverse-phase high-performance liquid chromatography, we purified hK11 to homogeneity. In seminal plasma, hK11 is present as a free enzyme of approximately 40 kDa. About 40% of hK11 is enzymatically active, whereas the rest is inactivated by internal cleavage after Arg156 (Genbank accession no. AF164623), which generates two peptides of approximately 20 kDa, connected by internal disulfide bonds. Purified hK11 possesses trypsin-like activity and cleaves synthetic peptides after arginine but not lysine residues. It does not cleave chymotrypsin substrates. Antithrombin, alpha1-antichymotrypsin, alpha2-antiplasmin, and alpha1-antitrypsin have no effect on hK11 activity and do not form complexes with hK11 in vitro. The strongest inhibitor, APMSF, completely inhibited hK11 activity at a concentration of 2.5 mmol/L. Aprotinin and an hK11-specific monoclonal antibody inhibited hK11 activity up to 40%. Plasmin is a strong candidate for cleaving hK11 at Arg156. This is the first report on purification and characterization of native hK11. We speculate that hK11, along with other kallikreins, proteases, and inhibitors, participates in a cascade enzymatic pathway responsible for semen liquefaction after ejaculation.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Trypsin inhibitor–Agarose, saline suspension, protein from Glycine max (soybean)