Merck
  • A new access to 3-substituted-1(2H)-isoquinolone by tandem palladium-catalyzed intramolecular aminocarbonylation annulation.
A new access to 3-substituted-1(2H)-isoquinolone by tandem palladium-catalyzed intramolecular aminocarbonylation annulation.
Organic & biomolecular chemistry (2012-03-01)
Antoine Dieudonné-Vatran, Michel Azoulay, Jean-Claude Florent
ABSTRACT
An original tribromide derivative based, palladium-catalyzed synthesis of 3-substituted-1(2H)-isoquinolone is described based on a regioselective Suzuki-Miyaura C-C coupling on o-halo-(2,2-dihalovinyl)-benzene followed by a palladium catalyzed amination-carbonylation-cyclization reaction. This sequence efficiently proceeds to build up isoquinolone in fair to good yields over a one-pot 3-bond synthesis reaction.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
4-Carboxyphenylboronic acid
Sigma-Aldrich
4-(Trifluoromethyl)phenylboronic acid, ≥95.0%
Sigma-Aldrich
3-Pyridinylboronic acid, ≥95.0%
Sigma-Aldrich
4-Nitrophenylboronic acid, ≥95.0%