Merck
Pharmacological characterization of MDL 105,519, an NMDA receptor glycine site antagonist.
European journal of pharmacology (1997-04-04)
B M Baron, B L Harrison, J H Kehne, C J Schmidt, P L van Giersbergen, H S White, B W Siegel, Y Senyah, T C McCloskey, G M Fadayel, V L Taylor, M K Murawsky, P Nyce, F G Salituro
ABSTRACT
MDL 105,519, (E)-3-(2-phenyl-2-carboxyethenyl)-4,6-dichloro-1 H-indole-2-carboxylic acid, is a potent and selective inhibitor of [3H]glycine binding to the NMDA receptor. MDL 105,519 inhibits NMDA (N-methyl-D-aspartate)-dependent responses including elevations of [3H]N-[1,(2-thienyl)cyclohexyl]-piperidine ([3H]TCP) binding in brain membranes, cyclic GMP accumulation in brain slices, and alterations in cytosolic CA2+ and NA(+)-CA2+ currents in cultured neurons. Inhibition was non-competitive with respect to NMDA and could be nullified with D-serine. Intravenously administered MDL 105,519 prevented harmaline-stimulated increases in cerebellar cyclic GMP content, providing biochemical evidence of NMDA receptor antagonism in vivo. This antagonism was associated with anticonvulsant activity in genetically based, chemically induced, and electrically mediated seizure models. Anxiolytic activity was observed in the rat separation-induced vocalization model, but muscle-relaxant activity was apparent at lower doses. Higher doses impair rotorod performance, but were without effect on mesolimbic dopamine turnover or prepulse inhibition of the startle reflex. This pattern of activities differentiates this compound from (5R,10S)-(+)-5-methyl-10, 11-dihydro-5H-dibenzo[a,d]cyclohepten-5,10-imine (MK-801) and indicates a lower psychotomimetic risk.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
MDL 105,519, ≥98% (HPLC), solid