Merck
Noradrenaline release in the rat subfornical organ area to blood pressure changes.
Experimental neurology (1998-08-26)
J Tanaka, Y Hayashi, T Watai, K Hori, M Nomura
ABSTRACT
To verify whether noradrenergic inputs to the subfornical organ (SFO) are involved in the control system of arterial pressure, we investigated the effects of blood pressure changes on noradrealine (NA) release in the SFO and its surrounding sites using microdialysis techniques in rats. Hemorrhage (5 or 10 ml/kg) significantly increased the NA concentration in the region of the SFO, but did not cause significant changes in the sites away from the SFO. An elevation in arterial pressure following intravenous administration of the alpha-agonist metaraminol slightly decreased the NA level in the region of the SFO. These results imply that the noradrenergic neural inputs to the SFO area may be involved in the control of cardiovascular function.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Metaraminol (+)-bitartrate salt