Merck
Affinity of muscarinic receptor antagonists for three putative muscarinic receptor binding sites.
British journal of pharmacology (1989-02-01)
R E Delmendo, A D Michel, R L Whiting
ABSTRACT
1. A range of muscarinic receptor antagonists were examined for affinity at the M1 muscarinic binding site, present in rat cerebrocortical membranes and the M2 muscarinic binding sites of rat cardiac and submaxillary gland membranes. 2. The results obtained were consistent with the presence of three classes of muscarinic binding site. 3. Both the M1 binding site, labelled by [3H]-pirenzepine ([3H]-Pir) in rat cerebrocortical membranes, and the M2 gland binding site, labelled by [3H]-N-methyl scopolamine ([3H]-NMS) in rat submaxillary gland membranes, displayed higher affinity for pirenzepine, dicyclomine, 4-diphenylacetoxy-N-methylpiperidine methiodide (4-DAMP) and cyclohexylphenyl (2-piperidinoethyl) silanol (CPPS) than did the M2 binding sites of cardiac membranes labelled by [3H]-NMS. 4. The M2 cardiac sites displayed higher affinity for methoctramine, himbacine and AF-DX 116 than did either the M1 binding site of cerebrocortical membranes or the M2 gland binding site present in rat submaxillary gland membranes. 5. The M1 and M2 gland binding sites could only be distinguished by considering the absolute affinity of compounds for these two sites. Thus, all compounds, with the exception of 4-DAMP, displayed between a 2 and 8 fold higher affinity for the M1 than for the M2 gland binding site. There were no antagonists with higher M2 gland than M1 affinity.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Methoctramine hydrate, ≥97% (NMR), solid