Merck
  • Estradiol-17β regulates the expression of insulin-like growth factors 1 and 2 via estradiol receptors in spotted scat (Scatophagus argus).
Estradiol-17β regulates the expression of insulin-like growth factors 1 and 2 via estradiol receptors in spotted scat (Scatophagus argus).
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology (2019-08-29)
Ke-Wei Zhang, Tian-Li Wu, Hua-Pu Chen, Dong-Neng Jiang, Chun-Hua Zhu, Si-Ping Deng, Yong Zhang, Guang-Li Li
ABSTRACT
Insulin-like growth factors (Igf1 and Igf2) play a key role in growth and development of vertebrates. In mammals, the expression of IGFs is regulated by estradiol-17β (E2) via estrogen receptors (ESRs). The expression of igfs can also be regulated by E2 in fish, while comparative study of this is still lacking. The present study examined tissue distribution of igfs and hepatic expression of igfs and esrs during gonad development in Scatophagus argus by real-time PCR. Serum E2 concentration was measured by enzyme-linked immunosorbent assay (ELISA). The hepatic expression of igfs and esrs at gonadal phase III, incubated with either E2 (0.1, 1 or 10 μM) alone or in combination with estrogen receptor antagonists-fulvestrant, MPP or PHTPP, was measured. igf1 and igf2 expressed highest in liver of both sexes. Igf1, esr1 and esr2b expressions and serum E2 concentration increased, while igf2 and esr2a expressions decreased, during ovary development. Igfs and esrs expressions increased while serum E2 concentration maintained low during testis development. In females, E2 incubation enhanced the expressions of igf1 and esr1 but inhibited that of igf2 and esr2a. Both fulvestrant and MPP inhibited up-regulation effect of E2 on igf1 and esr1. Fulvestrant enhanced down-regulation effect of E2 on igf2 and esr2a, but MPP conversely. In males, E2 incubation enhanced the expressions of igfs, esr1 and esr2a. Fulvestrant and MPP inhibited up-regulation effect of E2 on igfs and esr1. PHTPP inhibited igf1 and esr2 expressions in both sexes. Our results indicated that the expression of igfs is regulated by E2 via Esrs in S. argus.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
MPP dihydrochloride hydrate, ≥97% (HPLC), white, powder
Sigma-Aldrich
PHTPP, ≥98% (HPLC)