Merck
Regulation of endoplasmic reticulum turnover by selective autophagy.
Nature (2015-06-05)
Aliaksandr Khaminets, Theresa Heinrich, Muriel Mari, Paolo Grumati, Antje K Huebner, Masato Akutsu, Lutz Liebmann, Alexandra Stolz, Sandor Nietzsche, Nicole Koch, Mario Mauthe, Istvan Katona, Britta Qualmann, Joachim Weis, Fulvio Reggiori, Ingo Kurth, Christian A Hübner, Ivan Dikic
ABSTRACT
The endoplasmic reticulum (ER) is the largest intracellular endomembrane system, enabling protein and lipid synthesis, ion homeostasis, quality control of newly synthesized proteins and organelle communication. Constant ER turnover and modulation is needed to meet different cellular requirements and autophagy has an important role in this process. However, its underlying regulatory mechanisms remain unexplained. Here we show that members of the FAM134 reticulon protein family are ER-resident receptors that bind to autophagy modifiers LC3 and GABARAP, and facilitate ER degradation by autophagy ('ER-phagy'). Downregulation of FAM134B protein in human cells causes an expansion of the ER, while FAM134B overexpression results in ER fragmentation and lysosomal degradation. Mutant FAM134B proteins that cause sensory neuropathy in humans are unable to act as ER-phagy receptors. Consistently, disruption of Fam134b in mice causes expansion of the ER, inhibits ER turnover, sensitizes cells to stress-induced apoptotic cell death and leads to degeneration of sensory neurons. Therefore, selective ER-phagy via FAM134 proteins is indispensable for mammalian cell homeostasis and controls ER morphology and turnover in mice and humans.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Reagente per trasfezione GeneJuice®, Non-lipid based chemical transfection reagent optimized for maximum transfection efficiency, ease-of-use, and minimal cytotoxicity on a wide variety of mammalian cells.
Sigma-Aldrich
Anti-FAM134B (lumenal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-FAM134B antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution