Merck
  • VIP and PACAP potentiation of nicotinic ACh-evoked currents in rat parasympathetic neurons is mediated by G-protein activation.
VIP and PACAP potentiation of nicotinic ACh-evoked currents in rat parasympathetic neurons is mediated by G-protein activation.
The European journal of neuroscience (2000-08-18)
D M Liu, J Cuevas, D J Adams
ABSTRACT
The effects of vasoactive intestinal polypeptide (VIP) and pituitary adenylate cyclase-activating polypeptide (PACAP27 and PACAP38) on isolated parasympathetic neurons of rat intracardiac and submandibular ganglia were examined under voltage clamp using whole-cell patch-clamp recording techniques. VIP and PACAP (</= 10 nM) selectively and reversibly increased the affinity of nicotinic acetylcholine receptor channels (nAChRs) for their agonists resulting in a potentiation of acetylcholine (ACh)-evoked whole-cell currents at low agonist concentrations. VIP-induced potentiation was observed with either ACh or nicotine as the cholinergic agonist. The VIP- but not the PACAP-induced potentiation of ACh-evoked currents was inhibited by [Ac-Tyr1, D-Phe2]-GRF 1-29, amide (100 nM), a selective antagonist of VPAC1 and VPAC2 receptors; whereas the PACAP38- but not the VIP-induced potentiation was inhibited by 100 nM PACAP6-38, a PAC1 and VPAC2 receptor antagonist. The signal transduction pathway mediating VIP- and PACAP-induced potentiation of nicotinic ACh-evoked currents involves a pertussis toxin (PTX)-sensitive G-protein. Intracellular application of 200 microM GTPgammaS or GDPbetaS inhibited VIP-induced potentiation of ACh-evoked whole-cell currents. GTPgammaS alone potentiated ACh- and nicotine-evoked currents and the magnitude of these currents was not further increased by VIP or PACAP. The G-protein subtype modulating the neuronal nAChRs was examined by intracellular dialysis with antibodies directed against alphao, alphai-1,2, alphai-3 or beta G-protein subunits. Only the anti-Galphao and anti-Gbeta antibodies significantly inhibited the effect of VIP and PACAP on ACh-evoked currents. The potentiation of ACh-evoked currents by VIP and PACAP may be mediated by a membrane-delimited signal transduction cascade involving the PTX-sensitive Go protein.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
[Ac-Tyr1,D-Phe2]-VIP Antagonist-GRF 1-29