Merck
Novel antioxidant agents deriving from molecular combination of Vitamin C and NO-donor moieties.
Bioorganic & medicinal chemistry (2008-03-25)
Clara Cena, Konstantin Chegaev, Silvia Balbo, Loretta Lazzarato, Barbara Rolando, Marta Giorgis, Elisabetta Marini, Roberta Fruttero, Alberto Gasco
ABSTRACT
In this paper, we describe a new class of products in which NO-donor moieties are linked to either the 3-OH (4a-f) or 2-OH group (7a-c) of ascorbic acid (ASA). Log Ps and pK(a)s of these products were experimentally evaluated. All the compounds were tested for their antioxidant activity on lipidic peroxidation induced by Fe(3+)-ADP/NADPH in lipids of microsomal membranes of rat hepatocytes. Only 3-O series displays antioxidant activity and it seems to be principally dependent on the lipophilicity. Both series trigger in vitro NO-dependent vasodilator properties.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Acido L-ascorbico, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Acido L-ascorbico, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Acido L-ascorbico, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Acido L-ascorbico, 99%
Sigma-Aldrich
Acido L-ascorbico, reagent grade, crystalline
Sigma-Aldrich
Acido L-ascorbico, ACS reagent, ≥99%
Supelco
Acido L-ascorbico, analytical standard
Sigma-Aldrich
Acido L-ascorbico, meets USP testing specifications
Sigma-Aldrich
Acido L-ascorbico, reagent grade
Sigma-Aldrich
Acido L-ascorbico, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Acido L-ascorbico, FCC, FG
Sigma-Aldrich
Acido L-ascorbico, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Acido L-ascorbico, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Acido L-ascorbico, tested according to Ph. Eur.