Merck
  • RamA confers multidrug resistance in Salmonella enterica via increased expression of acrB, which is inhibited by chlorpromazine.
RamA confers multidrug resistance in Salmonella enterica via increased expression of acrB, which is inhibited by chlorpromazine.
Antimicrobial agents and chemotherapy (2008-08-13)
Andrew M Bailey, Ian T Paulsen, Laura J V Piddock
ABSTRACT
Salmonella enterica serovar Typhimurium SL1344, in which efflux pump genes (acrB, acrD, acrF, tolC) or regulatory genes thereof (marA, soxS, ramA) were inactivated, was grown in the presence of 240 antimicrobial and nonantimicrobial agents in the Biolog Phenotype MicroArray. Mutants lacking tolC, acrB, and ramA grew significantly worse than other mutants in the presence of 48 agents (some of which have not previously been identified as substrates of AcrAB-TolC) and particularly poorly in the presence of phenothiazines, which are human antipsychotics. MIC testing revealed that the phenothiazine chlorpromazine had antimicrobial activity and synergized with common antibiotics against different Salmonella serovars and SL1344. Chlorpromazine increased the intracellular accumulation of ethidium bromide, which was ablated in mutants lacking acrB, suggesting an interaction with AcrB. High-level but not low-level overexpression of ramA increased the expression of acrB; conferred resistance to chloramphenicol, tetracycline, nalidixic acid, and triclosan and organic solvent tolerance; and increased the amount of ethidium bromide accumulated. Chlorpromazine induced the modest overproduction of ramA but repressed acrB. These data suggest that phenothiazines are not efflux pump inhibitors but influence gene expression, including that of acrB, which confers the synergy with antimicrobials observed.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Esano, suitable for HPLC, ≥97.0% (GC)
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Esano, ReagentPlus®, ≥99%
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Esano, suitable for HPLC, ≥95%
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Cicloesano, ACS reagent, ≥99%
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Cicloesano, suitable for HPLC, ≥99.9%
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Esano, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Esano, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Cicloesano, suitable for HPLC, ≥99.7%
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Crystal Violet, certified by the BSC
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Esano, Laboratory Reagent, ≥95%
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Cicloesano, Laboratory Reagent, ≥99.8%
Prezzi e disponibilità al momento non sono disponibili
Supelco
Esano, analytical standard
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Cicloesano, puriss. p.a., ACS reagent, ≥99.5% (GC)
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Cicloesano, anhydrous, 99.5%
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Bromuro di etidio, BioReagent, Molecular Biology, 500 μg/mL in H2O
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Ethidium bromide, BioReagent, Molecular Biology, powder
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Crystal Violet Solution
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Crystal Violet, suitable for microscopy
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Nalidixic acid, ≥98%
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Acridine Orange solution, 2% in H2O
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Cicloesano, ACS reagent, ≥99%
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Crystal Violet Solution
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Bromuro di etidio, suitable for fluorescence, ~1% in H2O
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Crystal Violet, S. No.: 785
Prezzi e disponibilità al momento non sono disponibili
Supelco
Cicloesano, analytical standard
Prezzi e disponibilità al momento non sono disponibili