Merck
  • Discovery of novel lipophilic inhibitors of OXA-10 enzyme (class D beta-lactamase) by screening amino analogs and homologs of citrate and isocitrate.
Discovery of novel lipophilic inhibitors of OXA-10 enzyme (class D beta-lactamase) by screening amino analogs and homologs of citrate and isocitrate.
Bioorganic & medicinal chemistry letters (2009-05-27)
Joséphine Beck, Lionel Vercheval, Carine Bebrone, Adriana Herteg-Fernea, Patricia Lassaux, Jacqueline Marchand-Brynaert
ABSTRACT
Aminocitrate (and homolog) derivatives have been prepared by bis-alkylation of glycinate Schiff bases with bromoacetates (and ethyl acrylate), followed by N-acylation and esters (partial or complete) deprotection. Aminoisocitrate was similarly obtained by mono-alkylation with diethyl fumarate. Evaluation against representative beta-lactamases revealed that the free acid derivatives are modest inhibitors of class A enzymes, whilst their benzyl esters showed a good inhibition of OXA-10 (class D enzyme). A docking experiment featured hydrophobic interactions in the active site.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Acido citrico, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Acido citrico, ACS reagent, ≥99.5%
Sigma-Aldrich
Acido citrico, ≥99.5%, FCC, FG
Sigma-Aldrich
Acido citrico, 99%
Sigma-Aldrich
Acido citrico, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Acido citrico, BioUltra, anhydrous, ≥99.5% (T)