Merck
  • A novel fluorescent probe based on N, B, F co-doped carbon dots for highly selective and sensitive determination of sulfathiazole.
A novel fluorescent probe based on N, B, F co-doped carbon dots for highly selective and sensitive determination of sulfathiazole.
The Science of the total environment (2020-11-11)
Letian Chen, Yongli Liu, Guohao Cheng, Zegang Fan, Jinyu Yuan, Shuailong He, Guifen Zhu
ABSTRACT
The widespread occurrence of sulfathiazole (STZ) in the environment has raised concerns regarding the potential risks to ecosystem and human health. Thus, there is a need to develop facile and efficient methods for monitoring STZ. In this study, a novel fluorescent probe, based on N, B, F co-doped carbon dots (N, B, F-CDs), was developed for the highly sensitive and selective determination of STZ. The fluorescent N, B, F-CDs were prepared via a one-step hydrothermal method using malonate and 1-allyl-3-vinylimidazolium tetrafluoroborate ionic liquid as precursors. The obtained N, B, F-CDs exhibited excellent fluorescence response toward STZ due to the inner filter effect (IFE), which caused the fluorescence to be quenched. The fluorescent probe allowed the STZ concentration to be accurately determined with a low detection limit of 5.5 ng mL-1 in two wide linear ranges of 0.008-10 μg mL-1 and 10-45 μg mL-1. The practicability of the fluorescent probe was further validated in river water, soil, milk, and egg samples, and the satisfactory spiked recoveries of STZ ranged from 96.1 to 101.6%. The proposed fluorescent probe based on N, B, F-CDs can be easily prepared and possess high selectivity and sensitivity, thereby displaying its tremendous potential for the identification and determination of STZ in the environment.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
Sulfathiazole