Merck
  • Heterogeneity in radiation-induced DNA damage and repair in tumor and normal cells measured using the "comet" assay.
Heterogeneity in radiation-induced DNA damage and repair in tumor and normal cells measured using the "comet" assay.
Radiation Research (2012)
Olive PL, Banath JP, Durand RE