Merck
MKP1 promotes nonalcoholic steatohepatitis by suppressing AMPK activity through LKB1 nuclear retention.
Nature communications (2023-09-06)
Bin Qiu, Ahmed Lawan, Chrysovalantou E Xirouchaki, Jae-Sung Yi, Marie Robert, Lei Zhang, Wendy Brown, Carlos Fernández-Hernando, Xiaoyong Yang, Tony Tiganis, Anton M Bennett
ABSTRACT
Nonalcoholic steatohepatitis (NASH) is triggered by hepatocyte death through activation of caspase 6, as a result of decreased adenosine monophosphate (AMP)-activated protein kinase-alpha (AMPKα) activity. Increased hepatocellular death promotes inflammation which drives hepatic fibrosis. We show that the nuclear-localized mitogen-activated protein kinase (MAPK) phosphatase-1 (MKP1) is upregulated in NASH patients and in NASH diet fed male mice. The focus of this work is to investigate whether and how MKP1 is involved in the development of NASH. Under NASH conditions increased oxidative stress, induces MKP1 expression leading to nuclear p38 MAPK dephosphorylation and decreases liver kinase B1 (LKB1) phosphorylation at a site required to promote LKB1 nuclear exit. Hepatic deletion of MKP1 in NASH diet fed male mice releases nuclear LKB1 into the cytoplasm to activate AMPKα and prevents hepatocellular death, inflammation and NASH. Hence, nuclear-localized MKP1-p38 MAPK-LKB1 signaling is required to suppress AMPKα which triggers hepatocyte death and the development of NASH.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anticorpo monoclonale ANTI-FLAG® M2, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
N-acetil-L-cisteina, BioReagent, suitable for cell culture
Sigma-Aldrich
MitoTEMPO, ≥98% (HPLC)
Roche
Anti-HA-Biotin, High Affinity (3F10), from rat IgG1
Sigma-Aldrich
AMPK Inhibitor, Compound C, InSolution, ≥95%, 10 mM, AMPK Inhibitor