Merck
Actin architecture of cultured human thyroid cancer cells: predictor of differentiation?
Surgery (1990-12-01)
M J Demeure, M Hughes-Fulford, P E Goretzki, Q Y Duh, O H Clark
ABSTRACT
The actin cytoskeleton is important for cell structure and motility. A disordered actin architecture has been correlated with a high metastatic potential in melanoma, fibrosarcoma, and colon cancer models. Thyrotropin is known to induce growth and differentiation in cultured thyroid cells, whereas the carcinogenic phorbol ester 12-O-tetradecanoylphorbol 13-acetate (TPA) causes dedifferentiation and malignant transformation in many cell lines. We therefore assessed the effect of thyrotropin and TPA on the actin architecture of FTC-133 human follicular thyroid cancer cells in continuous culture. Staining of filamentous actin with rhodamine phalloidin showed that 1 mU/ml or 30 mU/ml thyrotropin-induced actin polymerization was detectable at 1 hour but more notable at 24 hours. Similarly TPA (0.008 to 10 mumol/L) caused rapid actin fiber disruption and redistribution to the cell periphery. Secondary antibody staining for alpha-actinin, a protein that binds and crosslinks actin, was more prominent after treatment with thyrotropin but decreased after TPA. These findings indicate that the actin cytoskeleton has a dynamic response to trophic factors. Thyrotropin promoted actin polymerization, but TPA caused depolymerization. These effects may correlate with cellular alpha-actinin levels. Actin architecture may therefore reflect the state of differentiation of thyroid tumor cells.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
FTC-133 Cell Line human, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.