Merck
  • Real-time PCR assay for the discrimination and quantification of wheat and barley strains of Wheat dwarf virus.
Real-time PCR assay for the discrimination and quantification of wheat and barley strains of Wheat dwarf virus.
Virus genes (2011-12-17)
S Gadiou, J Ripl, B Jaňourová, J Jarošová, J K Kundu
ABSTRACT
A rapid method for detection, discrimination and quantification of wheat and barley strains of wheat dwarf virus (WDV) was successfully developed. The sensitivity of quantification of the wheat and barley strains of WDV ranged from an average of 1.2 × 10(7)-1.2 × 10(2) and from an average of 1.4 × 10(7)-1.4 × 10(4) copies of viral genome, respectively. These standard serial dilutions were applied to plant and vector tissues for virus titer calculations. Both strains of WDV were clearly discriminated by specific probes and melting curve analysis. Both TaqMan(®) and SYBR(®) Green technologies provided accurate and reliable methods for monitoring, detection, discrimination, and quantification of WDV.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
GenElute Plant Genomic DNA Miniprep Kit, sufficient for 10 purifications