- De novo genome project of Cupriavidus basilensis OR16.
De novo genome project of Cupriavidus basilensis OR16.
Journal of bacteriology (2012-03-31)
Mátyás Cserháti, Balázs Kriszt, Sándor Szoboszlay, Ákos Tóth, István Szabó, András Táncsics, István Nagy, Balázs Horváth, István Nagy, József Kukolya
PMID22461549
ABSTRACT
Here we report on the complete genome sequence of Cupriavidus basilensis OR16 NCAIM BO2487. The genome of strain OR16 contains 7,534 putative coding sequences, including a large set of xenobiotics-degrading genes and a unique glucose dehydrogenase gene that is absent from other Cupriavidus genomes.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto