- Immobilized lectin columns: useful tools for the fractionation and structural analysis of oligosaccharides.
Immobilized lectin columns: useful tools for the fractionation and structural analysis of oligosaccharides.
Journal of chromatography (1992-04-24)
A Kobata, T Endo
PMID1517308
ABSTRACT
Elucidation of the binding specificity of a concanavalin A-Sepharose column led to the possibility of the affinity chromatography of oligosaccharides and glycopeptides with the use of immobilized lectin columns. Subsequent addition of immobilized erythroagglutinating phytohaemagglutinin, Aleuria aurantia lectin, Datura stramonium agglutinin, Ricinus communis agglutinin and Allomyrina dichotoma agglutinin to the range of well characterized lectin columns has afforded a way to fractionate a mixture of N-linked oligosaccharides even to a single component.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), agarose conjugate, Type VI, saline suspension