Merck
  • Resveratrol inhibits IgE-mediated basophilic mast cell degranulation and passive cutaneous anaphylaxis in mice.
Resveratrol inhibits IgE-mediated basophilic mast cell degranulation and passive cutaneous anaphylaxis in mice.
The Journal of nutrition (2013-03-22)
Seon-Young Han, Ji-Young Bae, Sin-Hye Park, Yun-Ho Kim, Jung Han Yoon Park, Young-Hee Kang
ABSTRACT
Resveratrol is a phytoalexin abundantly found in red grape skin and is effective in antitumor and antiinflammation associated with immune responses. This study investigated whether resveratrol suppressed immunoglobulin (Ig)E-mediated allergic responses and passive cutaneous anaphylaxis (PCA) in rat RBL-2H3 mast cells and in BALB/c mice. The release of β-hexosaminidase and histamine was enhanced in mast cells sensitized with anti-dinitrophenyl (DNP)-IgE and subsequently stimulated by DNP-human serum albumin (HSA), indicative of mast cell degranulation. When mast cells were pretreated with nontoxic resveratrol at 1-25 μmol/L, such induction was dose dependently diminished. Spleen tyrosine kinase (Syk) and phospholipase Cγ (PLCγ) of sensitized mast cells were activated by stimulation with DNP-HSA antigen, which was dampened by ≥5 μmol/L resveratrol. The phosphorylation of protein kinase C (PKC)μ and PKCθ was attenuated by administering resveratrol to DNP-HSA-exposed mast cells, whereas quiescent PKCζ/λ in sensitized cells was dose-dependently activated by resveratrol. Male BALB/c mice were sensitized for 24 h with DNP-IgE and orally administered with resveratrol 1 h before the DNP-HSA challenge. The histamine concentration was enhanced in sensitized mice challenged to DNP-HSA, which was reversed by administration of 10 mg/kg resveratrol. Additionally, it encumbered the tissue activation of Syk, PLCγ, and PKCμ in antigen-exposed mice. Resveratrol decreased IgE-mediated PCA and alleviated allergic edema of mouse ear and dorsal skin. Mast cell degranulation and allergic inflammation, accompanying the induction of monocyte chemotactic protein-1 and macrophage inflammatory protein-2, were inhibited by supplementing resveratrol to antigen-challenged mice. Resveratrol inhibited mast cell-derived, immediate-type allergic reactions, and these responses of resveratrol suggest possible therapeutic strategies in preventing allergic inflammatory diseases.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Histamine, ≥97.0%
Sigma-Aldrich
Phospholipase C from Clostridium perfringens (C. welchii), Type I, lyophilized powder, 10-50 units/mg protein
Sigma-Aldrich
β-N-Acetylglucosaminidase from Canavalia ensiformis (Jack bean), ammonium sulfate suspension, ≥8 units/mg protein
Sigma-Aldrich
Phospholipase C from Bacillus cereus, ≥200 units/mg protein
Sigma-Aldrich
β-N-Acetylglucosaminidase from Streptococcus pneumoniae, recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
Phospholipase C from Clostridium perfringens (C. welchii), Type XIV, lyophilized powder, ≥150 units/mg protein