Merck
A graphene oxide-based nano-beacon for DNA phosphorylation analysis.
Chemical communications (Cambridge, England) (2010-11-26)
Wenhe Wu, Haiyan Hu, Fan Li, Lihua Wang, Jimin Gao, Jianxin Lu, Chunhai Fan
ABSTRACT
We designed a single-fluorophore-tagged hairpin-structured nano-beacon probe by using a superquencher, graphene oxide (GO), based on which a new method for the analysis of DNA phosphorylation detection was developed.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Polynucleotide Kinase from T4-infected Escherichia coli, 10 units/μL, buffered aqueous glycerol solution