Merck
The PDGF signaling pathway controls multiple steroid-producing lineages.
Genes & development (2008-12-06)
Jennifer Schmahl, Kamran Rizzolo, Philippe Soriano
ABSTRACT
The platelet-derived growth factor (PDGF) signaling pathway regulates numerous lineages of mesenchymal cell origin during development and in the adult. The transcriptional targets of this pathway have been shown to be required in several PDGF-dependent processes, but the roles of these targets in specific tissues is just beginning to be identified. In this study, we show that five different PDGF target genes are essential for male and/or female fertility. Mutations in each of these five different genes lead to defects in the steroid-producing cells in the testis and/or ovary and altered hormone production, suggesting that the PDGF pathway controls steroidogenesis through these genes in both sexes. Furthermore, conditional mutations of both PDGF receptors revealed a requirement in steroid-producing cells in multiple organs, including the testis, ovary, and adrenal cortex. Therefore, PDGF signaling may constitute a common mechanism in the control of multiple steroidogenic lineages.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
Paraformaldehyde, prilled, 95%
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
Paraformaldehyde, meets analytical specification of DAC, 95.0-100.5%
Sigma-Aldrich
Anticorpo anti-fosfo-istone H3 (Ser10) marcatore di mitosi, Upstate®, from rabbit
Sigma-Aldrich
FragEL DNA Fragmentation Detection Kit, Fluorescent - TdT Enzyme