Merck
  • Studies on a new proteolytic enzyme from Achromobacter lyticus M497-1. II. specificity and inhibition studies of Achromobacter protease I.
Studies on a new proteolytic enzyme from Achromobacter lyticus M497-1. II. specificity and inhibition studies of Achromobacter protease I.
Biochimica et biophysica acta (1981-07-24)
T Masaki, T Fujihashi, K Nakamura, M Soejima
ABSTRACT
The unique specificity of Achromobacter protease I for lysine residue was investigated using synthetic and natural substrates, i.e., lysine derivatives, arginine derivatives, lysine vasopressin, substance P, ACTH and insulin. The enzyme cleaved only the -Lys-X- bonds in the above substrates. The binding affinity of alkylamines as determined by Ki was much stronger than that of the corresponding alkylguanidines.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Achromopeptidase, partially purified powder, ≥20,000 units/mg solid