Merck
Cl- secretory effects of EBIO in the rabbit conjunctival epithelium.
American journal of physiology. Cell physiology (2005-02-11)
Lawrence J Alvarez, Aldo C Zamudio, Oscar A Candia
ABSTRACT
Experiments were conducted to determine whether the Cl- secretagogue, 1-ethyl-2-benzimidazolinone (EBIO), stimulates Cl- transport in the rabbit conjunctival epithelium. For this study, epithelia were isolated in an Ussing-type chamber under short-circuit conditions. The effects of EBIO on the short-circuit current (I(sc)) and transepithelial resistance (R(t)) were measured under physiological conditions, as well as in experiments with altered electrolyte concentrations. Addition of 0.5 mM EBIO to the apical bath stimulated the control I(sc) by 64% and reduced R(t) by 21% (P < 0.05; paired data). Under Cl(-)-free conditions, I(sc) stimulation using EBIO was markedly attenuated. In the presence of an apical-to-basolateral K+ gradient and permeabilization of the apical membrane, the majority of the I(sc) reflected the transcellular movement of K+ via basolateral K+ channels. Under these conditions, EBIO in combination with A23187 elicited nearly instantaneous 60-90% increases in I(sc) that were sensitive to the calmodulin antagonist calmidazolium and the K+ channel blocker tetraethyl ammonium. In the presence of an apical-to-basolateral Cl- gradient and nystatin permeabilization of the basolateral aspect, EBIO increased the Cl(-)-dependent I(sc), an effect prevented by the channel blocker glibenclamide (0.3 mM). The latter compound also was used to determine the proportion of EBIO-evoked unidirectional 36Cl- fluxes in the presence of the Cl- gradient that traversed the epithelium transcellularly. Overall, EBIO activated apical Cl- channels and basolateral K+ channels (presumably those that are Ca2+ dependent), thereby suggesting that this compound, or related derivatives, may be suitable as topical agents to stimulate fluid transport across the tissue in individuals with lacrimal gland deficiencies.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
1-EBIO, ≥98% (HPLC)