Merck
Lysozyme viscoelastic matrices in tetramethylurea/water media: a small angle X-ray scattering study.
Biophysical chemistry (2002-10-16)
Marcelo A da Silva, Rosângela Itri, Elizabeth P G Arêas
ABSTRACT
Semi-solid viscoelastic matrices produced out of lysozyme in organic/aqueous media [tetramethylurea (TMU)/water] were characterized by small angle X-ray scattering (SAXS). The scattering curves were modeled in their form and interference factors. Radii of gyration of scattering particles were found to undergo a dramatic increase from 14 A in water to approximately 44 A in the matrices. Average correlation distances d=155 A were consistently verified for the scattering particles in the matrices, irrespective of solvent composition (in the 0.6<or=w(TMU)<or=0.8 range), in contrast to what is observed in water (d=62 A). At w(TMU)=0.9, however, a slight increase in R(g) (to R(g)=49 A) leads to interdigitation with the apolar prevailing medium of the matrix leading to the loss of the interference effect. Low dimensionality derived from the modeling procedure could be taken to indicate mass fractal character of the unfolded species, although polydispersion of samples could also have some contribution to that result. Despite the very significant shape distortion of the unfolded protein forms in the matrices, they still retain considerable globular character, as indicated by the Kratky plots obtained. The morphological results obtained in this work are compatible with the dynamical behavior displayed by the systems.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Tetramethylurea, 99%
Prezzi e disponibilità al momento non sono disponibili