Merck
Human volunteer study with PGME: eye irritation during vapour exposure.
Toxicology letters (2003-04-05)
H H Emmen, H Muijser, J H E Arts, M K Prinsen
ABSTRACT
The objective of this study was to establish the possible occurrence of eye irritation and subjective symptoms in human volunteers exposed to propylene glycol monomethyl ether (PGME) vapour at concentrations of 0, 100 and 150 ppm. Testing was conducted in 12 healthy male volunteers using a repeated measures design. Each subject was exposed for 2.5 h to each of the three exposure conditions that were spaced 7 days apart. The exposure sequences were counterbalanced and the exposure to the test substance and the effect measurements were conducted in a double-blind fashion. During all exposure sessions, 20 ppm diethyl ether was used as a 'masking agent' for vapour exposure. Measurements of pre- and post exposure eye redness, corneal thickness, tear film break-up time, conjunctival epithelial damage, blinking frequency, and subjective ratings on discomfort were used to evaluate the possible irritating effects of PGME. The results indicated no significant treatment effects for any of the objective parameters. Results of the subjective ratings indicated very slight effects on the eyes in the 150 ppm PGME condition only. No significant effects of treatment were found for the remaining questions concerning the perceived intensity of the smell in the room, the (un)pleasantness of the smell, the perceived effects on the skin, effects on the throat, shivering, muscle aching, and intestinal cramps. In conclusion, the results of the present study indicated minimal subjective eye effects at 150 ppm only, and no impact on the objective measures of eye irritation at either of the two exposure levels. It was concluded that the no adverse effect concentration for eye irritation due to PGME vapour was at least 150 ppm.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
1-Methoxy-2-propanol, ≥99.5%
Supelco
1-Methoxy-2-propanol, analytical standard