Merck
Bis(4-chlorophenyl) sulfone (BCPS) in Swedish marine and fresh water wildlife-a screening study.
Environment international (2004-03-31)
Karin Norström, Anders Olsson, Mats Olsson, Ake Bergman
ABSTRACT
Bis(4-chlorophenyl) sulfone (BCPS) is a high production volume chemical (HPVC) applied in thermostable polymers. BCPS has been detected as an environmental contaminant both in Europe and in North America but it is still not a commonly studied pollutant. In this study, three Baltic Sea fish species; herring (Clupea harengus), salmon (Salmo salar) and perch (Perca fluviatilis) from the Swedish coast, and one inland fish species, arctic char (Salvelinus alpinus), were analysed to screen for the occurrence and distribution of BCPS. Salmon and arctic char, were sampled in the early 1970s as well as the late 1990s. Fish eating grey seal (Halichoerus gryphus) and guillemot (Uria aalge) from the Baltic Sea were included to screen for whether BCPS biomagnify or not. The representativeness of the analysed samples for studying bioaccumulation of environmental pollutants was compared through analysis of two well known persistent and bioaccumulating compounds, 2,2',4,4',5,5'-hexachlorobiphenyl (CB-153), and 4,4'-DDE. Pooled muscle and blubber samples based on 4-10 individuals were used for analysis, as well as individual samples of grey seal blubber. 2,4,4'-Trichlorodiphenyl sulfone, was synthesised and applied as an internal standard. BCPS was detected in all marine samples but in only one of the fresh water fish samples. The highest BCPS concentrations detected, 1600 and 1900 ng/g lipid weighet (l.w.), were found in muscle from Baltic guillemot. The results indicate that BCPS is bioaccumulated in both grey seal and guillemot, and that the guillemot has higher concentrations of BCPS than the grey seal (50-500 ng/g l.w.). The concentrations found in different species of fish from the Baltic Sea ranged between 15-37 ng/g l.w. and lower concentrations were found in freshwater species (n.d.-1.8 ng/g l.w.). The present study shows that BCPS is found in all investigated species of wildlife but, in most species, still at low concentrations. However, the guillemot has levels in the low ppm range indicating a considerable biomagnification of BCPS in this species.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Bis(4-chlorophenyl) sulfone, 98%
Prezzi e disponibilità al momento non sono disponibili