Merck
Expanding the repertoire of heterocycle ring pairs for programmable minor groove DNA recognition.
Journal of the American Chemical Society (2004-08-19)
Michael A Marques, Raymond M Doss, Shane Foister, Peter B Dervan
ABSTRACT
The discrimination of the four Watson-Crick base pairs by minor groove DNA-binding polyamides have been attributed to the specificity of three five-membered aromatic amino acid subunits, 1-methyl-1H-imidazole (Im), 1-methyl-1H-pyrrole (Py), and 3-hydroxy-1H-pyrrole (Hp) paired four different ways. The search for additional ring pairs that demonstrate DNA-sequence specificity has led us to a new class of 6-5 fused bicycle rings as minor groove recognition elements. The affinities and specificities of the hydroxybenzimidazole/pyrrole (Hz/Py) and hydroxybenzimidazole/benzimidazole (Hz/Bi) pairs for each of the respective Watson-Crick base pairs within the sequence context 5'-TGGXCA-3' (X = A, T, G, C) were measured by quantitative DNaseI footprinting titrations. The Hz/Py and Hz/Bi distinguish T.A from A.T. Hairpin polyamides containing multiple Hz/Py pairs were examined and were shown to mimic the Hp/Py pair with regard to affinity and specificity. Therefore, the Hz/Py pair may be considered a second-generation replacement for the Hp/Py pair.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
N-Methylpyrrole, 99%