- A coronary artery fistula with saccular aneurysm mimicking a right atrial cystic mass.
A coronary artery fistula with saccular aneurysm mimicking a right atrial cystic mass.
Japanese heart journal (2004-09-09)
Byoung-Joo Choi, Hyuk-Jae Chang, So-Yeon Choi, Tae-Young Choi, Jo-Won Jung, Jaehoon Chung, Myeong-Ho Yoon, Gyo-Seung Hwang, Joon-Han Shin, Seung-Jea Tahk, Byung-Il William Choi
PMID15353882
ABSTRACT
A 16 year-old Korean girl with cardiomegaly was found to have a "right atrial cystic mass" on transthoracic echocardiography. An unusual cystic structure made the diagnosis difficult. However, transesophageal echocardiography and multidetector computed tomography revealed a coronary artery fistula with a distal saccular aneurysm involving the interatrial septum and draining into the right atrium. Multidetector computed tomography provided clear anatomic visualization that fully delineated the abnormal structures. Multidetector computed tomography may be considered as a good alternative for transesophageal echocardiography in assessing a tortuous coronary artery fistula and aneurysm with complex anatomy.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Poly(Ala, Glu, Lys, Tyr) 6:2:5:1 hydrobromide, Ala:Glu:Lys:Tyr (6:2:5:1), mol wt 20,000-30,000