Merck
  • Determination of impurities in pesticides and their degradation products formed during the wine-making process by solid-phase extraction and gas chromatography with detection by electron ionization mass spectrometry. II. Bromopropylate, trichlorphon, parathion-methyl and tebuconazole.
Determination of impurities in pesticides and their degradation products formed during the wine-making process by solid-phase extraction and gas chromatography with detection by electron ionization mass spectrometry. II. Bromopropylate, trichlorphon, parathion-methyl and tebuconazole.
Rapid communications in mass spectrometry : RCM (2004-10-14)
Juan José Jiménez, José Luis Bernal, M Jesús del Nozal, Laura Toribio, José Bernal
ABSTRACT
The presence of degradation products of bromopropylate, trichlorphon, parathion-methyl and tebuconazole in white and red wines elaborated from musts spiked with commercial formulations of the pesticides was studied. Must and wine were subjected to solid-phase extraction followed by gas chromatography with electron ionization mass spectrometric detection. Alpha-bromophenylphenylmethanol, aminoparathion, acetylaminoparathion-oxon and dichlorvos have been identified as degradation products of bromopropylate, parathion-methyl and trichlorphon in wines, respectively. Moreover, the presence of additives and impurities of the formulations in elaborated wines has also been found.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
Bromopropylate, PESTANAL®, analytical standard