Merck
Determining pirimiphos-methyl in durum wheat samples using an acetylcholinesterase inhibition assay.
Analytical and bioanalytical chemistry (2005-03-05)
Michele Del Carlo, Alessia Pepe, Marcello Mascini, Miriam De Gregorio, Angelo Visconti, Dario Compagnone
ABSTRACT
An electrochemical assay used for detecting acetylcholinesterase (AChE) inhibitors has been optimised to detect pirimiphos-methyl in durum wheat. Pirimiphos-methyl is a phosphothionate insecticide and so it needs to be transformed into the corresponding oxo form to act as an effective AChE inhibitor. The inhibition assay was based on the electrochemical detection of the product of AChE, choline, via choline oxidase biosensors obtained with Prussian-Blue modified screen printed electrodes. The procedure for the oxidation of pirimiphos methyl via N-bromosuccinimide (NBS) and AChE inhibition was optimised for reagent concentrations and inhibition time in a buffer solution. A calibration of the pirimiphos-methyl (25-1,000 ng/ml) was obtained in the buffer. The intra-electrode CV ranged between 1.6 and 15.0, whereas the inter-electrode CV ranged between 4.6 and 16.0. The detection limit (LOD) was 38 ng/ml, and the I(50%) was 360 ng/ml. The assay conditions were then re-optimised to work with durum wheat extracts, and the calibrations were obtained under different experimental conditions, such as sample pretreatment (milled or whole grains) and extract concentration. The calibrations were slightly affected by the sample matrix, resulting in an increased LOD (65-133 ng/ml) and I(50%) (640-1,650 ng/ml). The LOD found for the sample, determined under optimal conditions, was 3 mg/kg. Spiked samples were prepared at the EU regulated level (5 mg/kg) and analysed with the optimised protocol, resulting in an average recovery of 70.3%.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
Pirimiphos-methyl, PESTANAL®, analytical standard