Merck
  • [Determination of residual epichlorohydrin and sym-dichloroisopropyl alcohol in cationic etherified reagent by gas chromatography].
[Determination of residual epichlorohydrin and sym-dichloroisopropyl alcohol in cationic etherified reagent by gas chromatography].
Se pu = Chinese journal of chromatography (2005-04-06)
Weiyi Zhou, Guoliang Fan, Dongfeng Jiang
ABSTRACT
A method for the determination of the residual epichlorohydrin and sym-dichloroisopropyl alcohol in cationic etherified reagent by gas chromatography has been established. Methyl benzoate, which has high extraction efficiency for the two components, was used as extractant. With an HP-5 capillary column, the two components were baseline separated and they eluted before the extractant. The linear ranges achieved were 5 - 590 mg/kg for epichlorohydrin and 21 - 480 mg/kg for sym-dichloroisopropyl alcohol. The limits of detection were 1.2 mg/kg for epichlorohydrin and 2.2 mg/kg for sym-dichloroisopropyl alcohol. Recoveries for epichlorohydrin were 95.93% - 103.42% with relative standard deviations of 2.4% - 10.6% and those for sym-dichloroisopropyl alcohol were 98.54% - 107.40% with relative standard deviations of 6.6% -11.1%. The method is simple, fast, and convenient.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
1,3-Dichloro-2-propanol, 97%