Merck
Continuous biotreatment of copper-concentrated solutions by biosorption with Sargassum sp.
Applied biochemistry and biotechnology (2006-01-18)
Francisca Pessôa de França, Flávia Pinto Padilha, Antonio Carlos Augusto da Costa
ABSTRACT
Seaweed Sargassum sp. biomass proved to be useful for the recovery of ionic copper from highly concentrated solutions simulating effluents from semiconductor production. In the case of solutions containing copper in the form of chloride, sulfate, and nitrate salts, the best pH for the recovery of copper was 4.5. It was observed that copper biosorption from copper nitrate solutions was higher than the recovery of copper from copper chloride or copper sulfate solutions. The continuous system used was constituted of four column reactors filled with the biomass of Sargassum sp. and showed high operational stability. The biomass of Sargassum sp. in the reactors was gradually saturated from the bottom to the top of each column reactor. The biomass of Sargassum sp. in the first column saturated first, followed by a gradual saturation of the remaining columns owing to preconcentration performed by the biomass in the first column. The biomass of Sargassum in the bioreactors completely biosorbed the ionic copper contained in 63 L of copper sulfate solution, 72 L of copper chloride solution, and 72 L of copper nitrate solution, all the solutions containing copper at 500 mg/L. Effluents produced after biosorption presented copper concentrations < 0.5 mg/L.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Copper(II) hydroxide, technical grade