Merck
  • Arginase activity in a murine macrophage cell line (RAW264.7) stimulated with lipopolysaccharide from Actinobacillus actinomycetemcomitans.
Arginase activity in a murine macrophage cell line (RAW264.7) stimulated with lipopolysaccharide from Actinobacillus actinomycetemcomitans.
Oral microbiology and immunology (2006-04-22)
W Sosroseno, M Musa, M Ravichandran, M Fikri Ibrahim, P S Bird, G J Seymour
ABSTRACT
The aim of the present study was to determine whether or not lipopolysaccharide from Actinobacillus actinomycetemcomitans could stimulate arginase activity in a murine macrophage cell line (RAW264.7 cells). RAW264.7 cells were treated with A. actinomycetemcomitans-lipopolysaccharide or lipopolysaccharide from Escherichia coli for 24 h. The effect of polymyxin B, l-norvaline, dl-norvaline, dexamethasone and cytokines (interferon-gamma and interleukin-4) on arginase activity in A. actinomycetemcomitans-lipopolysaccharide-stimulated cells was also determined. The cells were pretreated with anti-CD14, anti -toll-like receptor 2, or anti-toll-like receptor 4 antibody prior to stimulation with A. actinomycetemcomitans-lipopolysaccharide. Arginase activity was determined by a colorimetric assay. A. actinomycetemcomitans-lipopolysaccharide stimulated arginase activity in RAW264.7 cells in a dose-dependent manner, but was less potent than E. coli-lipopolysaccharide. Polymyxin B and l-norvaline, but not dl-norvaline, blocked the arginase activity in A. actinomycetemcomitans-lipopolysaccharide-stimulated cells. Dexamethasone and interleukin-4 but not interferon-gamma augmented arginase activity in A. actinomycetemcomitans-lipopolysaccharide-stimulated cells. Treatment of the cells with anti-CD14 and anti-toll-like receptor 4 but not anti-toll-like receptor 2 antibody decreased arginase activity in A. actinomycetemcomitans-lipopolysaccharide-stimulated cells. The results of the present study suggest that lipopolysaccharide from A. actinomycetemcomitans via CD14/toll-like receptor 4 complex molecules and the regulatory control of glucocorticoid and cytokines may stimulate arginase activity in RAW264.7 cells.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
L-Norvaline, arginase inhibitor
Sigma-Aldrich
DL-Norvaline