Merck
Asymmetric synthesis of cyclopropylamines starting from N-sulfinyl alpha-chloro ketimines.
Organic letters (2007-01-16)
Bram Denolf, Sven Mangelinckx, Karl W Törnroos, Norbert De Kimpe
ABSTRACT
Treatment of novel chiral N-sulfinyl alpha-chloro ketimines with Grignard reagents resulted in the synthesis of chiral N-(1-substituted cyclopropyl)-tert-butanesulfinamides in acceptable to good yields and diastereoselectivity via 1,3-dehydrohalogenation and subsequent addition of the Grignard reagent to the intermediate cyclopropylideneamine. Only in the case of allylmagnesium chloride did the reaction lead to aziridines in high yield. Further deprotection toward N-unprotected 1-substituted cyclopropylamines was established, and the absolute configuration was determined. [reaction: see text].
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Cyclopropylamine, 98%