Merck
  • Ligand accelerated indium(III)-catalyzed asymmetric alkynylation of aldehydes with 2-methyl-3-butyn-2-ol as an ethyne equivalent donor.
Ligand accelerated indium(III)-catalyzed asymmetric alkynylation of aldehydes with 2-methyl-3-butyn-2-ol as an ethyne equivalent donor.
Chemical communications (Cambridge, England) (2007-02-22)
Shinji Harada, Ryo Takita, Takashi Ohshima, Shigeki Matsunaga, Masakatsu Shibasaki
ABSTRACT
Indium(III)-catalyzed asymmetric alkynylation of aryl, heteroaryl, alkyl and alkenyl aldehydes with 2-methyl-3-butyn-2-ol as an ethyne equivalent donor was realized, and products were obtained in moderate to good yields (up to 97%) and high enantioselectivities (up to 99% ee) using 2-10 mol% of catalyst.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
2-metil-3-butin-2-olo, 98%