Merck
Two phases of calcium requirement during starfish meiotic maturation.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology (2007-02-24)
Hiroaki Tosuji, Yukari Seki, Keiichiro Kyozuka
ABSTRACT
During meiosis in oocytes of the starfish, Asterina pectinifera, a Ca(2+) transient has been observed. To clarify the role of Ca(2+) during oocyte maturation in starfish, an intracellular Ca(2+) blocker, TMB-8, was applied. The oocyte maturation induced by 1-methyladenine (1-MA) was blocked by 100 microM TMB-8. Reinitiation of meiosis with germinal vesicle breakdown (GVBD) and the following chromosome condensation did not take place. Maturation-promoting factor (MPF) activity did not increase and GVBD and chromosome condensation did not occur. Ca(2+) transient observed immediately after 1-MA application in control oocytes was also blocked by TMB-8. When calyculin A, which activate the MPF directly, was applied to the oocytes instead of 1-MA in seawater containing 100 microM TMB-8, GVBD and chromosome condensation were blocked. Cytoplasmic transplantation studies confirmed that MPF was activated, although TMB-8 blocked GVBD. These results show that TMB-8 blocked the increase of MPF activity induced by 1-MA and the process of active MPF inducing GVBD and subsequent chromosome condensation. Together with the above phenomena, it is conceivable that there are two phases of Ca(2+) requirement during starfish oocyte maturation. These are the activation of MPF, moreover, GVBD, and the subsequent chromosome condensation.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
8-(Diethylamino)octyl-3,4,5-trimethoxybenzoate hydrochloride, ≥98%