Merck
Fragrance material review on alpha-ionone.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2007-11-27)
J Lalko, A Lapczynski, V T Politano, D McGinty, S Bhatia, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of alpha-ionone when used as a fragrance ingredient is presented.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
α-Ionone, ≥90%, stabilized
Sigma-Aldrich
α-Ionone, technical grade, 90%