Merck
Solid-phase organic synthesis of polyisoprenoid alcohols with traceless sulfone linker.
The Journal of organic chemistry (2008-08-19)
Yi-Fan Chang, Chen-Yu Liu, Chih-Wei Guo, Yen-Chih Wang, Jim-Min Fang, Wei-Chieh Cheng
ABSTRACT
Solid-phase organic synthesis of polyprenols with a traceless sulfone linker is described. The polymer-bound benezenesulfinate is first linked with the "tail" building blocks of isoprenyl chlorides via S-alkylation. With use of dimsyl anion as an appropriate base, the polymer-bound alpha-sulfonyl carbanion is generated and coupled with other "body" building blocks in an efficient manner. After repeated processes and a global palladium-catalyzed desulfonation with LiEt 3BH as the reducing agent, the desired polyprenols with various chain lengths and geometrical configurations are obtained in 32-59% overall yields. The solid-phase synthesis offers the advantage in facile isolation of polyprenols without tedious operation or time-consuming purification.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
3-Methyl-2-buten-1-ol, 99%
Sigma-Aldrich
3-Methyl-2-buten-1-ol, ≥98%, FG