Merck
Pomegranate juice inhibits sulfoconjugation in Caco-2 human colon carcinoma cells.
Journal of medicinal food (2008-12-05)
Ayako Saruwatari, Shigeaki Okamura, Yoko Nakajima, Yuji Narukawa, Tadahiro Takeda, Hiroomi Tamura
ABSTRACT
Several fruit juices have been reported to cause food-drug interactions, mainly affecting cytochrome P450 activity; however, little is known about the effects of fruit juices on conjugation reactions. Among several fruit juices tested (apple, peach, orange, pineapple, grapefruit, and pomegranate), pomegranate juice potently inhibited the sulfoconjugation of 1-naphthol in Caco-2 cells. This inhibition was both dose- and culture time-dependent, with a 50% inhibitory concentration (IC(50)) value calculated at 2.7% (vol/vol). In contrast, no obvious inhibition of glucuronidation of 1-naphthol in Caco-2 cells was observed by any of the juices examined. Punicalagin, the most abundant antioxidant polyphenol in pomegranate juice, was also found to strongly inhibit sulfoconjugation in Caco-2 cells with an IC(50) of 45 microM, which is consistent with that of pomegranate juice. These data suggest that punicalagin is mainly responsible for the inhibition of sulfoconjugation by pomegranate juice. We additionally demonstrated that pomegranate juice and punicalagin both inhibit phenol sulfotransferase activity in Caco-2 cells in vitro, at concentrations that are almost equivalent to those used in the Caco-2 cells. Pomegranate juice, however, shows no effects on the expression of the sulfotransferase SULT1A family of genes (SULT1A1 and SULT1A3) in Caco-2 cells. These results indicate that the inhibition of sulfotransferase activity by punicalagin in Caco-2 cells is responsible for the reductions seen in 1-naphthyl sulfate accumulation. Our data also suggest that constituents of pomegranate juice, most probably punicalagin, impair the enteric functions of sulfoconjugation and that this might have effects upon the bioavailability of drugs and other compounds present in food and in the environment. These effects might be related to the anticarcinogenic properties of pomegranate juice.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Punicalagin, ≥98% (HPLC), from pomegranate
Supelco
Punicalagin, analytical standard