Merck
  • Screening, quantification, and confirmation of phenylbutazone and oxyphenbutazone in equine plasma by liquid chromatography-tandem mass spectrometry.
Screening, quantification, and confirmation of phenylbutazone and oxyphenbutazone in equine plasma by liquid chromatography-tandem mass spectrometry.
Journal of analytical toxicology (2009-01-24)
Youwen You, Cornelius E Uboh, Lawrence R Soma, Fuyu Guan, Xiaoqing Li, Jeffrey A Rudy, Jinwen Chen
ABSTRACT
A sensitive liquid chromatographic-tandem mass spectrometric method was developed and validated for screening, quantification, and confirmation of phenylbutazone and oxyphenbutazone in equine plasma. Analytes were recovered from plasma by liquid-liquid extraction followed by separation in a reversed-phase column and identification by mass spectrometry with selected reaction monitoring in negative electrospray ionization mode. Extraction recovery for both analytes was >80%. Limits of detection, quantification, and confirmation for both analytes were 0.01 microg/mL (S/N>or= 3), 0.05 microg/mL, and 0.05 microg/mL, respectively. The assay with d9-labeled phenylbutazone as internal standard (IS) was linear over a range of 0.05-20 microg/mL (r2>0.995). Intra- and interday precision in terms of coefficient of variation was less than 15%. Intra- and interday accuracy (bias%) was within 80-120%. Hemolysis of red blood cells decreased analyte signal intensity but did not affect quantification results because an isotope-labeled IS was used. Analytes were stable in plasma for 24 h at room temperature, 9 days at 4 degrees C, and 45 days at -20 degrees C and -70 degrees C. The method was successfully used in screening, quantification, and confirmation of phenylbutazone in post-competition plasma samples obtained from racehorses. The method is simple, rapid, and reliably reproducible.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Oxyphenbutazone, ≥98% (HPLC)