- Acetylcholinesterase and butyrylcholinesterase inhibition of ethanolic extract and monoterpenes from Pimpinella anisoides V Brig. (Apiaceae).
Acetylcholinesterase and butyrylcholinesterase inhibition of ethanolic extract and monoterpenes from Pimpinella anisoides V Brig. (Apiaceae).
Fitoterapia (2009-04-09)
Federica Menichini, Rosa Tundis, Monica R Loizzo, Marco Bonesi, Mariangela Marrelli, Giancarlo A Statti, Francesco Menichini, Filomena Conforti
PMID19351555
ABSTRACT
Ethanolic extract from the fruits of Pimpinella anisoides, an aromatic plant and a spice, exhibited activity against AChE and BChE, with IC(50) values of 227.5 and 362.1 microg/ml, respectively. The most abundant constituents of the extract were trans-anethole, (+)-limonene and (+)-sabinene. trans-Anethole exhibited the highest activity against AChE and BChE with IC(50) values of 134.7 and 209.6 microg/ml, respectively. The bicyclic monoterpene (+)-sabinene exhibited a promising activity against AChE (IC(50) of 176.5 microg/ml) and BChE (IC(50) of 218.6 microg/ml).
MATERIALI