Merck
Influence of soil properties on the enantioselective dissipation of the herbicide lactofen in soils.
Journal of agricultural and food chemistry (2009-06-11)
Jinling Diao, Chunguang Lv, Xinquan Wang, Ziheng Dang, Wentao Zhu, Zhiqiang Zhou
ABSTRACT
A scheme was developed to elucidate the dissipation behaviors of the two enantiomers of the herbicide lactofen in soils using a normal-phase high-performance liquid chromatograph (HPLC) with UV detector and a column with a cellulose-tri-(3,5-dimethylphenylcarbamate)-based chiral stationary phase (CDMPC-CSP). Eight soils with a wide range of soil properties were studied. Racemic and the enantiopure (S)-(+)- and (R)-(-)-lactofen were incubated under aerobic and anaerobic conditions. The data from sterilized controls indicated that the dissipation of lactofen was biological. The dissipation was shown to be enantioselective with (S)-(+)-enantiomer being degraded faster than the (R)-(-)-enantiomer, resulting in residues enriched with (R)-(-)-lactofen when the racemic compound was incubated. Lactofen was configurationally stable in soil, showing no interconversion of (S)-(+)- to (R)-(-)- enantiomer and vice versa. Significant correlations of the enantioselectivity, expressed as ES = (k((S)) - k((R)))/(k((S)) + k((R))) of lactofen with soil pH were observed under aerobic and anaerobic conditions. In addition, we found that the enantioselectivity correlated with the soil texture rather than the organic carbon.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
Lactofen, PESTANAL®, analytical standard