Merck
  • Allosteric modulators of the adenosine A1 receptor: synthesis and pharmacological evaluation of 4-substituted 2-amino-3-benzoylthiophenes.
Allosteric modulators of the adenosine A1 receptor: synthesis and pharmacological evaluation of 4-substituted 2-amino-3-benzoylthiophenes.
Journal of medicinal chemistry (2009-06-12)
Luigi Aurelio, Celine Valant, Bernard L Flynn, Patrick M Sexton, Arthur Christopoulos, Peter J Scammells
ABSTRACT
A series of 4-substituted 2-amino-3-benzoylthiophenes was screened using a functional assay of A(1)AR-mediated phosphorylation of ERK1/2 in intact CHO cells to identify both potential agonistic effects as well the ability to allosterically modulate the activity of the orthosteric agonist, R-PIA. More detailed concentration-response experiments were subsequently performed on two compounds (9a and 9o) utilizing both the ERK1/2 assay as well as a second assay of [(35)S]GTPgammaS binding to activated G proteins.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
(−)-N6-(2-Phenylisopropyl)adenosine, solid